missing translation for 'onlineSavingsMsg'
Learn More

D-Panthenol, 98+%

Catalog No. AAA1849922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1849922 100 g
AAA1849936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1849922 Supplier Thermo Scientific Chemicals Supplier No. A1849922

CAS: 81-13-0 | C9H19NO4 | 205.25 g/mol

D-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.

Solubility
Fully miscible in water and soluble in alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.

Chemical Identifiers

CAS 81-13-0
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00065006
InChI Key SNPLKNRPJHDVJA-ZETCQYMHSA-N
Synonym dexpanthenol, d-panthenol, pantothenol, bepanthen, d-pantothenyl alcohol, ilopan, +-panthenol, bepanthene, panthoderm, thenalton
PubChem CID 131204
ChEBI CHEBI:27373
IUPAC Name (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCCO

Specifications

Density 1.162
Boiling Point 118°C to 120°C (0.02 mmHg)
Flash Point 159°C (318°F)
Refractive Index 1.499
Quantity 100 g
Beilstein 1724947
Sensitivity Hygroscopic
Merck Index 14,2947
Solubility Information Fully miscible in water and soluble in alcohol.
Optical Rotation −30° (c=5 in Water)
Formula Weight 205.26
Percent Purity ≥98%
Chemical Name or Material D-Panthenol
Show More Show Less
EINECSNumber 201-327-3
RTECSNumber ES4316000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only