missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Panthenol, 98+%

Catalog No. AC232805000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232805000 500 g Glass bottle
AC232801000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232805000 Supplier Thermo Scientific Chemicals Supplier No. 232805000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 81-13-0 | C9H19NO4 | 205.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81-13-0
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00065006
InChI Key SNPLKNRPJHDVJA-ZETCQYMHSA-N
Synonym dexpanthenol, d-panthenol, pantothenol, bepanthen, d-pantothenyl alcohol, ilopan, +-panthenol, bepanthene, panthoderm, thenalton
PubChem CID 131204
ChEBI CHEBI:27373
IUPAC Name (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCCO

Specifications

Color Colorless to Yellow
Boiling Point 118°C to 120°C (2.7 Pa)
Infrared Spectrum Authentic
Assay Percent Range 98% min
Packaging Glass bottle
Refractive Index 1.4950 to 1.5020
Linear Formula HOCH2C(CH3)2CH(OH)CONH(CH2)3OH
Quantity 500 g
Beilstein 04,III,75
Merck Index 15, 2949
Specific Rotation Condition +30.50° (20°C c=5,H2O on anh. sub)
Specific Rotation + 30.50
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in methanol,ethanol,sparingly soluble in chloroform,slightly soluble in diethyl ether,insoluble in fats and oils
Formula Weight 205.26
Percent Purity 98+%
Physical Form Viscous Liquid or Semi Solid
Chemical Name or Material D-Panthenol, 98+%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 201-327-3
RTECSNumber ES4316000
TSCA TSCA

RUO – Research Use Only