missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Pantethine, 98%, Spectrum™ Chemical
P1024, 16816-67-4, C22H42N4O8S2
$345.76 - $345.76
Chemical Identifiers
| CAS | 16816-67-4 |
|---|---|
| Molecular Formula | C22H42N4O8S2 |
| Molecular Weight (g/mol) | 554.72 |
| InChI Key | DJWYOLJPSHDSAL-ROUUACIJSA-N |
| IUPAC Name | (2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide |
| SMILES | CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO |
Description
Spectrum™ Chemical D-Pantethine , also known as Bis-pantethine, can be used to treat acne and improve the blood-cholesterol profile. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 16816-67-4 | |
| 554.72 | |
| (2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide |
| C22H42N4O8S2 | |
| DJWYOLJPSHDSAL-ROUUACIJSA-N | |
| CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO |
Specifications
| 16816-67-4 | |
| Amber Glass Bottle | |
| 1 g | |
| CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO | |
| 554.72 | |
| Reagent |
| 100% | |
| C22H42N4O8S2 | |
| DJWYOLJPSHDSAL-ROUUACIJSA-N | |
| (2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide | |
| 98% |