missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Norvaline 99.0+%, TCI America™
$180.97 - $1622.92
Chemical Identifiers
| CAS | 2013-12-9 |
|---|---|
| Molecular Formula | C5H11NO2 |
| Molecular Weight (g/mol) | 117.15 |
| MDL Number | MFCD00008097 |
| InChI Key | SNDPXSYFESPGGJ-SCSAIBSYSA-N |
| Synonym | d-norvaline, d--norvaline, 2r-2-aminopentanoic acid, h-d-nva-oh, d---norvaline, d-2-aminovaleric acid, d-2-aminopentanoic acid, r-2-aminovaleric acid, r-2-aminopentanoic acid, d-ape |
| PubChem CID | 439575 |
| ChEBI | CHEBI:28804 |
| IUPAC Name | (2R)-2-aminopentanoic acid |
| SMILES | CCC[C@@H](N)C(O)=O |
Chemical Identifiers
| 2013-12-9 | |
| 117.15 | |
| SNDPXSYFESPGGJ-SCSAIBSYSA-N | |
| 439575 | |
| (2R)-2-aminopentanoic acid |
| C5H11NO2 | |
| MFCD00008097 | |
| d-norvaline, d--norvaline, 2r-2-aminopentanoic acid, h-d-nva-oh, d---norvaline, d-2-aminovaleric acid, d-2-aminopentanoic acid, r-2-aminovaleric acid, r-2-aminopentanoic acid, d-ape | |
| CHEBI:28804 | |
| CCC[C@@H](N)C(O)=O |
Specifications
| 2013-12-9 | |
| C5H11NO2 | |
| 1 g | |
| SNDPXSYFESPGGJ-SCSAIBSYSA-N | |
| (2R)-2-aminopentanoic acid | |
| 439575 | |
| 117.15 | |
| Crystalline Powder |
| White | |
| MFCD00008097 | |
| d-norvaline, d--norvaline, 2r-2-aminopentanoic acid, h-d-nva-oh, d---norvaline, d-2-aminovaleric acid, d-2-aminopentanoic acid, r-2-aminovaleric acid, r-2-aminopentanoic acid, d-ape | |
| CCC[C@@H](N)C(O)=O | |
| 117.15 | |
| CHEBI:28804 | |
| ≥99.0% (T) | |
| D-Norvaline |
Safety and Handling
TSCA : No