missing translation for 'onlineSavingsMsg'
Learn More

D-Norvaline 99.0+%, TCI America™

Catalog No. N06735G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06735G 5 g
N06731G 1 g
N067325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N06735G Supplier TCI America Supplier No. N06735G
Only null left

Chemical Identifiers

CAS 2013-12-9
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00008097
InChI Key SNDPXSYFESPGGJ-SCSAIBSYSA-N
Synonym d-norvaline, d--norvaline, 2r-2-aminopentanoic acid, h-d-nva-oh, d---norvaline, d-2-aminovaleric acid, d-2-aminopentanoic acid, r-2-aminovaleric acid, r-2-aminopentanoic acid, d-ape
PubChem CID 439575
ChEBI CHEBI:28804
IUPAC Name (2R)-2-aminopentanoic acid
SMILES CCC[C@@H](N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 117.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Norvaline