missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Norvaline, 99%

Catalog No. AAB2344403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2344403 1 g
AAB2344406 5 g
AAB2344414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2344403 Supplier Thermo Scientific Chemicals Supplier No. B2344403
Only null left

CAS: 2013-12-9 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2013-12-9
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00008097
InChI Key SNDPXSYFESPGGJ-SCSAIBSYSA-N
Synonym d-norvaline, d--norvaline, 2r-2-aminopentanoic acid, h-d-nva-oh, d---norvaline, d-2-aminovaleric acid, d-2-aminopentanoic acid, r-2-aminovaleric acid, r-2-aminopentanoic acid, d-ape
PubChem CID 439575
ChEBI CHEBI:28804
IUPAC Name (2R)-2-aminopentanoic acid
SMILES CCC[C@@H](N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721161
Merck Index 14,6716
Quantity 1 g
Formula Weight 117.15
Optical Rotation −25° (c=10 in 20% HCl)
Percent Purity 99%
Chemical Name or Material D-Norvaline
EINECSNumber 217-936-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only