missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Melibiose Monohydrate 99.0+%, TCI America™

Catalog No. p-7131951
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00501G 1 g
M005010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M00501G Supplier TCI America Supplier No. M00501G

Chemical Identifiers

CAS 66009-10-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00198188
InChI Key CHIDEFLSUMQFBY-UHFFFAOYNA-N
Synonym d +-melibiose monohydrate, d-+-melibiose hydrate, 585-99-9 anhydrous, melibiose hplc, 6-, a-d-galactopyranosyl-d-glucopyranose, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci
PubChem CID 71308738
IUPAC Name 2,3,4,5-tetrahydroxy-6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O

Specifications

Melting Point 185°C
Color White
Quantity 1 g
Formula Weight 342.30
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Melibiose Monohydrate