missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Melibiose monohydrate, 99+%

CAS: 66009-10-7 | C12H24O12 | 360.31 g/mol

$539.41 - $539.41

Chemical Identifiers

CAS 66009-10-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00198188
InChI Key CHIDEFLSUMQFBY-UHFFFAOYNA-N
Synonym d +-melibiose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci, d-+-melibiose hydrate, 585-99-9 anhydrous, 6-, 6-, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate
PubChem CID 71308738
IUPAC Name (2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
SMILES O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125370250
SDS View Documents
Thermo Scientific Chemicals
125370250
25 g Glass bottle
Each for $539.41
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

66009-10-7
360.31
CHIDEFLSUMQFBY-UHFFFAOYNA-N
71308738
O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O
C12H24O12
MFCD00198188
d +-melibiose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci, d-+-melibiose hydrate, 585-99-9 anhydrous, 6-, 6-, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate
(2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate

Specifications

66009-10-7
182.0°C
Authentic
Odorless
C12H24O12
25 g
14,582
+ 135.00
Solubility in water: soluble
O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O
360.31
360.32
Crystalline Powder
0.4mL 0.1N NaOH/6g max.
White
99+%
Glass bottle
MFCD00198188
31,421
+ 135.00 (20.00°C c=4,H2O/NH3)
d +-melibiose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci, d-+-melibiose hydrate, 585-99-9 anhydrous, 6-, 6-, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate
CHIDEFLSUMQFBY-UHFFFAOYNA-N
(2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
71308738
99+%
D(+)-Melibiose monohydrate
Videos
SDS
Documents

Documents

RUO – Research Use Only