missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Melibiose monohydrate, 99+%

Catalog No. AC125375000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125375000 500 g Glass bottle
AC125370250 25 g Glass bottle
AC125371000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125375000 Supplier Thermo Scientific Chemicals Supplier No. 125375000
Only null left

CAS: 66009-10-7 | C12H24O12 | 360.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66009-10-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00198188
InChI Key CHIDEFLSUMQFBY-UHFFFAOYNA-N
Synonym d +-melibiose monohydrate, d-+-melibiose hydrate, 585-99-9 anhydrous, melibiose hplc, 6-, a-d-galactopyranosyl-d-glucopyranose, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci
PubChem CID 71308738
IUPAC Name (2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
SMILES O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O

Specifications

Acidity 0.4mL 0.1N NaOH/6g max.
Melting Point 182.0°C
Color White
Infrared Spectrum Authentic
Odor Odorless
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 500 g
Beilstein 31,421
Merck Index 14,582
Specific Rotation Condition + 135.00 (20.00°C c=4,H2O/NH3)
Specific Rotation + 135.00
Solubility Information Solubility in water: soluble
Formula Weight 360.32
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Melibiose monohydrate
Show More Show Less

RUO – Research Use Only