Learn More
Thermo Scientific Chemicals D-(+)-Melezitose hydrate, 99%
CAS: 207511-10-2 | C18H32O16 | 504.44 g/mol
$109.45 - $344.23
Chemical Identifiers
| CAS | 207511-10-2 |
|---|---|
| Molecular Formula | C18H32O16 |
| Molecular Weight (g/mol) | 504.44 |
| MDL Number | MFCD00006627 |
| InChI Key | QWIZNVHXZXRPDR-IXUUBQBONA-N |
| Synonym | d-+-melezitose hydrate |
| PubChem CID | 126963632 |
| IUPAC Name | (2R,3S,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;hydrate |
| SMILES | OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2220909
|
Thermo Scientific Chemicals
B2220909 |
10 g |
N/A
|
|
|||||
|
AAB2220918
|
Thermo Scientific Chemicals
B2220918 |
50 g |
N/A
|
|
|||||
Description
D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents,water, moisture.
Chemical Identifiers
| 207511-10-2 | |
| 504.44 | |
| QWIZNVHXZXRPDR-IXUUBQBONA-N | |
| 126963632 | |
| OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O |
| C18H32O16 | |
| MFCD00006627 | |
| d-+-melezitose hydrate | |
| (2R,3S,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;hydrate |
Specifications
| 207511-10-2 | |
| C18H32O16 | |
| 10 g | |
| Hygroscopic | |
| d-+-melezitose hydrate | |
| QWIZNVHXZXRPDR-IXUUBQBONA-N | |
| OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O | |
| 504.44 | |
| 504.46 (Anhydrous) | |
| D-(+)-Melezitose hydrate |
| ∼160°C (decomposition) | |
| MFCD00006627 | |
| 99539 | |
| 14,5819 | |
| Soluble in water. | |
| +88°(c=4 in Water) | |
| (2R,3S,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;hydrate | |
| 126963632 | |
| 99% |
Safety and Handling
EINECSNumber : 209-894-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only