missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Melezitose hydrate, 99%

Catalog No. AAB2220909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2220909 10 g
AAB2220918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2220909 Supplier Thermo Scientific Chemicals Supplier No. B2220909
Only null left

CAS: 207511-10-2 | C18H32O16 | 504.44 g/mol

D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents,water, moisture.

Chemical Identifiers

CAS 207511-10-2
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.44
MDL Number MFCD00006627
InChI Key QWIZNVHXZXRPDR-IXUUBQBONA-N
Synonym d-+-melezitose hydrate
PubChem CID 126963632
IUPAC Name (2R,3S,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;hydrate
SMILES OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O

Specifications

Melting Point ∼160°C (decomposition)
Quantity 10 g
Beilstein 99539
Sensitivity Hygroscopic
Merck Index 14,5819
Solubility Information Soluble in water.
Optical Rotation +88°(c=4 in Water)
Formula Weight 504.46 (Anhydrous)
Percent Purity 99%
Chemical Name or Material D-(+)-Melezitose hydrate
EINECSNumber 209-894-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only