accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Chemicals
Organic compounds
Unclassified Organic Compounds
44410050MG
Structure Search
Print
MilliporeSigma™ D-Mannose-6-phosphate, Disodium Salt, Calbiochem™,
Catalog No.
44410050MG
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
50 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
Packaging
44410050MG
50 mg
Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
44410050MG
Supplier
MilliporeSigma™
Supplier No.
44410050MG
Sign In
or
Register
to check your price and availability.
$174.79
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
33068-18-7
Molecular Formula
C6H13Na2O9P
Molecular Weight (g/mol)
306.114
InChI Key
UUYHWNKAOZTLOW-OXIHULNRSA-N
PubChem CID
131852496
IUPAC Name
sodium;[(2R,3R,4S,5S)-2,3,4,5-tetrahydroxy-6-oxohexyl] dihydrogen phosphate
SMILES
C(C(C(C(C(C=O)O)O)O)O)OP(=O)(O)O.[Na].[Na]
Specifications
Color
White
Packaging
Glass Bottle
Quantity
50 mg
Product Suggestions
Documents