missing translation for 'onlineSavingsMsg'
Learn More

D-Mannono-1,4-lactone 97.0+%, TCI America™

Catalog No. M09581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09581G 1 g
M09585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09581G Supplier TCI America Supplier No. M09581G

Chemical Identifiers

CAS 26301-79-1
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00065020
InChI Key SXZYCXMUPBBULW-SQOUGZDYSA-N
PubChem CID 1774581
IUPAC Name (3S,4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Melting Point 153°C
Color White
Quantity 1 g
Formula Weight 178.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-Mannono-1,4-lactone