missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Mandelic Acid 99.0+%, TCI America™

Supplier:  TCI America M0662100G

 View more versions of this product

Catalog No. M0662100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

D-(-)-Mandelic Acid
611-71-2
C8H8O3
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
O[C@@H](C(O)=O)C1=CC=CC=C1
152.15
CHEBI:17656
≥99.0% (GC,T)
Crystalline Powder
134°C
White to Yellow
MFCD00064251
IWYDHOAUDWTVEP-SSDOTTSWSA-N
(2R)-2-hydroxy-2-phenylacetic acid
11914
152.15
100 g

Chemical Identifiers

611-71-2
152.15
IWYDHOAUDWTVEP-SSDOTTSWSA-N
11914
(2R)-2-hydroxy-2-phenylacetic acid
C8H8O3
MFCD00064251
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
CHEBI:17656
O[C@@H](C(O)=O)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1764

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions