missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Mandelic Acid 99.0+%, TCI America™

Catalog No. M066225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M066225G 25 g
M0662100G 100 g
M0662500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M066225G Supplier TCI America Supplier No. M066225G

Chemical Identifiers

CAS 611-71-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064251
InChI Key IWYDHOAUDWTVEP-SSDOTTSWSA-N
Synonym r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
PubChem CID 11914
ChEBI CHEBI:17656
IUPAC Name (2R)-2-hydroxy-2-phenylacetic acid
SMILES O[C@@H](C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 134°C
Color White to Yellow
Formula Weight 152.15
Percent Purity ≥99.0% (GC,T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Mandelic Acid
EINECSNumber (3)-1764
TSCA Yes