missing translation for 'onlineSavingsMsg'
Learn More

D-Limonene, Food Grade, 95-97.5%, Spectrum™ Chemical

Catalog No. 18602287
Change view
Click to view available options
Quantity:
500 g
2.5 kg
15 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602287 500 g Amber Glass Bottle
18602288 2.5 kg Amber Glass Bottle
18602289 15 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602287 Supplier Spectrum Chemical Mfg Cor Supplier No. L1599500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

L1599, 5989-27-5, C10H16

Spectrum™ Chemical D-Limonene, Food Grade , also known as 4-Isopropenyl-1-methylcyclohexene, is used as a flavorant and as a fragrance in perfumery, aftershave lotions, bath products and other such products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 5989-27-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062991
InChI Key XMGQYMWWDOXHJM-JTQLQIEISA-N
IUPAC Name (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
SMILES CC(=C)[C@@H]1CCC(C)=CC1

Specifications

CAS Max % 100%
Boiling Point 175°C to 176°C
Melting Range -95.5°C to -74°C
Packaging Amber Glass Bottle
Refractive Index 1.471-1.474
Quantity 500 g
UN Number UN2052
Optical Rotation +96.0° to +104°
Percent Purity 95 to 97.5%
Grade Food Grade
Physical Form Liquid
Chemical Name or Material D-Limonene
Show More Show Less