missing translation for 'onlineSavingsMsg'
Learn More

D-Limonene, 96.9%, MP Biomedicals

Catalog No. ICN15523480
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15523480 100 mL
ICN15523490 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15523480 Supplier MP Biomedicals Inc Supplier No. 0215523480

Chemical Identifiers

CAS 5989-27-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062991
InChI Key XMGQYMWWDOXHJM-JTQLQIEISA-N
Synonym d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
PubChem CID 440917
ChEBI CHEBI:15382
SMILES CC(=C)[C@@H]1CCC(C)=CC1

Specifications

Color Colorless to Yellow
Density ∽0.84 g/mL
Quantity 100 mL
IUPAC Name (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Formula Weight 136.26
Percent Purity 0.969
Physical Form Liquid
Chemical Name or Material D-Limonene