missing translation for 'onlineSavingsMsg'
Learn More

D-Leucine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. L01985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01985G 5 g
L01981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. L01985G missing translation for 'mfr' TCI America missing translation for 'supplierNo' L01985G

Chemical Identifiers

CAS 5845-53-4
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00066115
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
Synonym d-leucine methyl ester hydrochloride, r-methyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-ome.hcl, d-leucine, methyl ester, hydrochloride, methyl 2r-2-amino-4-methylpentanoate, chloride, h-d-leu-ome hcl, methyl d-leucinate hydrochloride, d-leu-och3.hcl, d-leucine,methyl ester, pubchem10893
PubChem CID 11600782
IUPAC Name methyl (2R)-2-amino-4-methylpentanoate hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC(C)C

Specifications

Melting Point 151°C
Color White
Quantity 5 g
Formula Weight 181.66
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Leucine Methyl Ester Hydrochloride