missing translation for 'onlineSavingsMsg'
Learn More

D-Leucine methyl ester hydrochloride, 98%

Catalog No. p-7056506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6545606 5 g
AAH6545614 25 g
AAH6545622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6545606 Supplier Thermo Scientific Chemicals Supplier No. H6545606

CAS: 5845-53-4 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5845-53-4
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00066115
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
Synonym d-leucine methyl ester hydrochloride, r-methyl 2-amino-4-methylpentanoate hydrochloride, h-d-leu-ome.hcl, d-leucine, methyl ester, hydrochloride, methyl 2r-2-amino-4-methylpentanoate, chloride, h-d-leu-ome hcl, methyl d-leucinate hydrochloride, d-leu-och3.hcl, d-leucine,methyl ester, pubchem10893
PubChem CID 11600782
IUPAC Name methyl (2R)-2-amino-4-methylpentanoate hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC(C)C

Specifications

Melting Point 151°C
Color White
Beilstein 3655228
Quantity 5 g
Formula Weight 181.66
Percent Purity 98%
Sensitivity Hygroscopic
Physical Form Solid
Chemical Name or Material D-Leucine methyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only