missing translation for 'onlineSavingsMsg'
Learn More

D-Lactose Monohydrate (Powder/NF/EP/BP/JP), Fisher Chemical™

$3027.38 - $3027.38

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
MDL Number MFCD00150747
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
L8-12
SDS View Documents
Fisher Chemical
L812
12 kg Poly Pail
Each for $3,027.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

64044-51-5
360.312
WSVLPVUVIUVCRA-ZEAJSSQNSA-N
133126686
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
C12H24O12
MFCD00150747
d-lactose monohydrate, d-+-lactosemonohydrate
(3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate

Specifications

219°C
64044-51-5
12 kg
MFCD00150747
WSVLPVUVIUVCRA-ZEAJSSQNSA-N
(3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
133126686
BP/EP/JP/NF
0.5% max. (JP/NF)
Pass Test (BP/EP/JP/NF)
>219°C
D-Lactose Monohydrate
White
Solid
C12H24O12
d-lactose monohydrate, d-+-lactosemonohydrate
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
360.312
360.32
Pass Test (EP/BP/NF)
Poly Pail
Negligible
1.525g/cm³
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'