Learn More
Thermo Scientific Chemicals D-Isoleucine, 98%
CAS: 319-78-8 | C6H13NO2 | 131.18 g/mol
Supplier: Thermo Scientific Chemicals H2748803
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 265°C (sublimation) | |
| 319-78-8 | |
| MFCD00064221 | |
| AGPKZVBTJJNPAG-LXDRQGDMNA-N | |
| (2R,3R)-2-amino-3-methylpentanoic acid | |
| 131.18 | |
| CHEBI:27730 | |
| 131.18 |
| D-Isoleucine | |
| C6H13NO2 | |
| d-isoleucine, 2r,3r-2-amino-3-methylpentanoic acid, h-d-ile-oh, isoleucine, d, unii-v87gja0g54, r-isoleucine, 2r,3r-2-amino-3-methylvaleric acid, alloisoleucin, d-ile, ambotzhaa1186 | |
| CC[C@@H](C)[C@@H](N)C(O)=O | |
| 1 g | |
| 76551 | |
| −40° (c=5 in 5N HCl) | |
| 98% |
Chemical Identifiers
| 319-78-8 | |
| 131.18 | |
| AGPKZVBTJJNPAG-LXDRQGDMNA-N | |
| 76551 | |
| (2R,3R)-2-amino-3-methylpentanoic acid |
| C6H13NO2 | |
| MFCD00064221 | |
| d-isoleucine, 2r,3r-2-amino-3-methylpentanoic acid, h-d-ile-oh, isoleucine, d, unii-v87gja0g54, r-isoleucine, 2r,3r-2-amino-3-methylvaleric acid, alloisoleucin, d-ile, ambotzhaa1186 | |
| CHEBI:27730 | |
| CC[C@@H](C)[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : 206-269-2
RTECSNumber : NR4700000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only