missing translation for 'onlineSavingsMsg'
Learn More

D-Homophenylalanine 98.0+%, TCI America™

$33.56 - $126.76

Chemical Identifiers

CAS 82795-51-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00063091
InChI Key JTTHKOPSMAVJFE-SECBINFHSA-N
Synonym d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
PubChem CID 6950302
IUPAC Name (2R)-2-amino-4-phenylbutanoic acid
SMILES N[C@H](CCC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0984100MG
View Documents
TCI America
H0984100MG
100 mg
Each for $33.56
Only null left
 
H09841G
View Documents
TCI America
H09841G
1 g
Each for $126.76
Only null left
 
Description

Description

Specifications

Chemical Identifiers

82795-51-5
179.22
JTTHKOPSMAVJFE-SECBINFHSA-N
6950302
N[C@H](CCC1=CC=CC=C1)C(O)=O
C10H13NO2
MFCD00063091
d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
(2R)-2-amino-4-phenylbutanoic acid

Specifications

82795-51-5
C10H13NO2
100 mg
JTTHKOPSMAVJFE-SECBINFHSA-N
(2R)-2-amino-4-phenylbutanoic acid
6950302
≥98.0% (T)
D-Homophenylalanine
White-Yellow
MFCD00063091
d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
N[C@H](CCC1=CC=CC=C1)C(O)=O
179.22
179.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'