missing translation for 'onlineSavingsMsg'
Learn More

D-Homophenylalanine 98.0+%, TCI America™

Catalog No. H0984100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0984100MG 100 mg
H09841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0984100MG Supplier TCI America Supplier No. H0984100MG

Chemical Identifiers

CAS 82795-51-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00063091
InChI Key JTTHKOPSMAVJFE-SECBINFHSA-N
Synonym d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
PubChem CID 6950302
IUPAC Name (2R)-2-amino-4-phenylbutanoic acid
SMILES N[C@H](CCC1=CC=CC=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 179.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Homophenylalanine