missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Homophenylalanine, 98%
CAS: 82795-51-5 | C10H13NO2 | 179.22 g/mol
$140.86 - $471.34
Chemical Identifiers
| CAS | 82795-51-5 |
|---|---|
| Molecular Formula | C10H13NO2 |
| Molecular Weight (g/mol) | 179.22 |
| MDL Number | MFCD00063091 |
| InChI Key | JTTHKOPSMAVJFE-SECBINFHSA-N |
| Synonym | d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine |
| PubChem CID | 6950302 |
| IUPAC Name | (2R)-2-amino-4-phenylbutanoic acid |
| SMILES | N[C@H](CCC1=CC=CC=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2762903
|
Thermo Scientific Chemicals
H2762903 |
1 g |
Each for $140.86
|
|
|||||
|
AAH2762906
|
Thermo Scientific Chemicals
H2762906 |
5 g |
Each for $471.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 82795-51-5 | |
| 179.22 | |
| JTTHKOPSMAVJFE-SECBINFHSA-N | |
| 6950302 | |
| N[C@H](CCC1=CC=CC=C1)C(O)=O |
| C10H13NO2 | |
| MFCD00063091 | |
| d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine | |
| (2R)-2-amino-4-phenylbutanoic acid |
Specifications
| >300°C | |
| C10H13NO2 | |
| 4675530 | |
| JTTHKOPSMAVJFE-SECBINFHSA-N | |
| (2R)-2-amino-4-phenylbutanoic acid | |
| 179.22 | |
| 179.22 | |
| D-Homophenylalanine |
| 82795-51-5 | |
| MFCD00063091 | |
| d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine | |
| N[C@H](CCC1=CC=CC=C1)C(O)=O | |
| 1 g | |
| 6950302 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only