missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamine 98.0+%, TCI America™

Catalog No. G02781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02781G 1 g
G02785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02781G Supplier TCI America Supplier No. G02781G
Only null left

Chemical Identifiers

CAS 5959-95-5
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00065607
InChI Key ZDXPYRJPNDTMRX-GSVOUGTGSA-N
Synonym d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln
PubChem CID 145815
ChEBI CHEBI:17061
IUPAC Name (2R)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Color White
Quantity 1 g
Formula Weight 146.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Glutamine