missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glutamine, 99+%

Catalog No. AAJ6078403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6078403 1 g
AAJ6078406 5 g
AAJ6078414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6078403 Supplier Thermo Scientific Chemicals Supplier No. J6078403
Only null left

Kinase inhibitor; has antivasospastic properties | CAS: 5959-95-5 | C5H10N2O3 | 146.146 g/mol

Kinase inhibitor; has antivasospastic properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5959-95-5
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00065607
InChI Key ZDXPYRJPNDTMRX-GSVOUGTGSA-N
Synonym d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln
PubChem CID 145815
ChEBI CHEBI:17061
IUPAC Name (2R)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Color White
Beilstein 1723796
Quantity 1 g
Formula Weight 146.14
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D-Glutamine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only