missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glutamic acid 1-tert-butyl ester, 98%
Description
D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Color | White |
| Solubility Information | Slightly soluble in water. |
| Quantity | 1 g |
| Formula Weight | 203.24 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | D-Glutamic acid 1-tert-butyl ester |
Safety and Handling
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only