missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamic acid 1-tert-butyl ester, 98%

Catalog No. AAH6331803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6331803 1 g
AAH6331806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6331803 Supplier Thermo Scientific Chemicals Supplier No. H6331803
Only null left

CAS: 25456-76-2 | C9H17NO4 | 203.238 g/mol

D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glutamic acid 1-tert-butyl ester, is an aminoacid which is widely used in pharmaceutical and food industry. It is also used as a pharmaceutical intermediate. It is also an important organic intermediate.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 25456-76-2
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD09842819
InChI Key QVAQMUAKTNUNLN-ZCFIWIBFSA-N
Synonym h-d-glu-otbu, r-4-amino-5-tert-butoxy-5-oxopentanoic acid, 4r-4-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamic acid 1-tert-butyl ester, r-4-amino-5-tert-butoxy-5-oxopentanoicacid
PubChem CID 7019726
IUPAC Name (4R)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(=O)O)N

Specifications

Color White
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 203.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Glutamic acid 1-tert-butyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only