missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamic acid 1-benzyl ester, 95%

Catalog No. p-7056779
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6590306 5 g
AAH6590314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH6590306 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H6590306

CAS: 79338-14-0 | C12H15NO4 | 237.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79338-14-0
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00237065
InChI Key HFZKKJHBHCZXTQ-SNVBAGLBSA-N
Synonym h-d-glu-obzl, 1-benzyl d-glutamate, r-4-amino-5-benzyloxy-5-oxopentanoic acid, d-glutamic acid 1-benzyl ester, 7-benzyl glutamate, 4r-4-amino-5-benzyloxy-5-oxopentanoic acid
PubChem CID 7019728
IUPAC Name (4R)-4-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)N

Specifications

Color White
Beilstein 3208599
Quantity 5 g
Formula Weight 237.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material D-Glutamic acid 1-benzyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only