missing translation for 'onlineSavingsMsg'
Learn More

D-Glucurono-6,3-lactone 99.0+%, TCI America™

Catalog No. p-7130118
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G005525G 25 g
G0055100G 100 g
G0055500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G005525G Supplier TCI America Supplier No. G005525G
Only null left

Chemical Identifiers

CAS 32449-92-6
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.124
MDL Number MFCD00135622
InChI Key UYUXSRADSPPKRZ-KKQCNMDGSA-N
Synonym d-glucuro-3,6-lactone
PubChem CID 91634076
IUPAC Name (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
SMILES C(=O)C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 178°C
Color White
pH 3.5 (10% H2O soln.)
Quantity 25 g
Formula Weight 176.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucurono-6,3-lactone
EINECSNumber (9)-0437
RTECSNumber LZ8930000
TSCA Yes