missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glucurono-3,6-lactone, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 500 g |
| Molecular Weight (g/mol) | 176.13 |
| Grade | Ungraded |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | D-Glucurono-3,6-lactone |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.