missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic Acid 96.0+%, TCI America™

Supplier:  TCI America G03025G

 View more versions of this product

Catalog No. G03025G


Only null left

Description

Description

Specifications

Specifications

D-Glucuronic Acid
153°C
C6H10O7
5 g
IAJILQKETJEXLJ-QTBDOELSSA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
65041
194.14
Crystalline Powder
6556-12-3
White-Yellow
MFCD00077778
2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
C(=O)C(C(C(C(C(=O)O)O)O)O)O
194.14
CHEBI:47953
≥96.0% (HPLC,T)

Chemical Identifiers

6556-12-3
194.14
IAJILQKETJEXLJ-QTBDOELSSA-N
65041
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
C6H10O7
MFCD00077778
2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
CHEBI:47953
C(=O)C(C(C(C(C(=O)O)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0522

RTECSNumber : LZ8836600

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions