missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic acid, 98%

Catalog No. AC309880100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309880100 10 g Plastic bottle
AC309880250 25 g Plastic bottle
AC309881000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC309880100 Supplier Thermo Scientific Chemicals Supplier No. 309880100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6556-12-3 | C6H10O7 | 194.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00064368
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
ChEBI CHEBI:47953
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

Specifications

Melting Point 158.0°C to 162.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 10 g
Merck Index 15,4501
Specific Rotation Condition + 36.50 (20.00°C c=6,H2O,after 2 hrs)
Specific Rotation + 36.50
Synonym 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
SMILES C(=O)C(C(C(C(C(=O)O)O)O)O)O
PubChem CID 65041
Formula Weight 194.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D-Glucuronic acid, 98%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 229-486-4
TSCA TSCA

RUO – Research Use Only