missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronamide 98.0+%, TCI America™

Catalog No. g022325g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G022325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G022325G Supplier TCI America Supplier No. G022325G
Only null left

Chemical Identifiers

CAS 3789-97-7
Molecular Formula C6H11NO6
Molecular Weight (g/mol) 193.155
MDL Number MFCD00006619
InChI Key VOIFKEWOFUNPBN-UHFFFAOYSA-N
Synonym glucuronamide, d-glucuronic acid amide, voifkewofunpbn-uhfffaoysa, 3,4,5,6-tetrahydroxytetrahydro-2h-pyran-2-carboxamide non-preferred name
PubChem CID 3482
IUPAC Name 3,4,5,6-tetrahydroxyoxane-2-carboxamide
SMILES C1(C(C(OC(C1O)O)C(=O)N)O)O

Specifications

Melting Point 167°C
Quantity 25 g
Formula Weight 193.16
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucuronamide
EINECSNumber (9)-0339
RTECSNumber LZ8829200
TSCA No