missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronamide, 98%

Catalog No. AAJ6604706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6604706 5 g
AAJ6604703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6604706 Supplier Thermo Scientific Chemicals Supplier No. J6604706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3789-97-7 | C6H11NO6 | 193.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3789-97-7
Molecular Formula C6H11NO6
Molecular Weight (g/mol) 193.155
MDL Number MFCD00006619
InChI Key VOIFKEWOFUNPBN-UHFFFAOYSA-N
Synonym glucuronamide, d-glucuronic acid amide, voifkewofunpbn-uhfffaoysa, 3,4,5,6-tetrahydroxytetrahydro-2h-pyran-2-carboxamide non-preferred name
PubChem CID 3482
IUPAC Name 3,4,5,6-tetrahydroxyoxane-2-carboxamide
SMILES C1(C(C(OC(C1O)O)C(=O)N)O)O

Specifications

Melting Point 166°C to 167°C
Color Tan to White
Quantity 5 g
Solubility Information Soluble in water.
Optical Rotation +35° (c=2 in Water)
Formula Weight 193.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Glucuronamide
EINECSNumber 223-260-9
RTECSNumber LZ8829200
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only