missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Glucose monohydrate, 99+%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Ignition Residue | 0.1% max |
| Assay Percent Range | 99% min. (ELSD) (HPLC) |
| Packaging | Plastic Bottle |
| Quantity | 1 kg |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 198.17 |
| Percent Purity | 99+% |
| Grade | Extra Pure |
| Chemical Name or Material | D(+)-Glucose Monohydrate |
RUO – Research Use Only