missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose monohydrate, 99%

CAS: 14431-43-7

$168.84 - $168.84

Chemical Identifiers

CAS 14431-43-7
MDL Number MFCD00149450
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA472630E
View Documents
Thermo Scientific Chemicals
0472630E
2.5 kg
Each for $168.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14431-43-7
MFCD00149450

Specifications

14431-43-7
White
2.5 kg
198.18 (180.16 Anhydrous)
Powder
∼83°C
MFCD00149450
3694181
99%
D-(+)-Glucose monohydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-075-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only