missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose monohydrate

Catalog No. AAA1109036
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1109036 500 g
AAA110900E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1109036 Supplier Thermo Scientific Chemicals Supplier No. A1109036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14431-43-7 | C6H12O6·H2O

D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O
MDL Number MFCD00149450

Specifications

Melting Point ∽83°C
Assay Percent Range 99%
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 198.18 (180.16anhydrous)
Chemical Name or Material D-(+)-Glucose monohydrate
EINECSNumber 200-075-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only