missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glucose 6-Phosphate Barium Salt Heptahydrate 98.0+%, TCI America™
$42.87 - $115.57
Chemical Identifiers
| CAS | 58823-95-3 |
|---|---|
| Molecular Formula | C6H13O9P |
| MDL Number | MFCD00221521 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G0052100MG
|
TCI America
G0052100MG |
100 mg |
Each for $42.87
|
|
|||||
|
G00521G
|
TCI America
G00521G |
1 g |
Each for $115.57
|
|
|||||
Chemical Identifiers
| 58823-95-3 | |
| C6H13O9P | |
| MFCD00221521 |
Specifications
| 58823-95-3 | |
| C6H13O9P | |
| 100 mg | |
| 395.45 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00221521 | |
| 1564 | |
| ≥98.0% (W) | |
| D-Glucose 6-Phosphate Barium Salt Heptahydrate |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator