missing translation for 'onlineSavingsMsg'
Learn More

D-Glucosamic Acid 98.0+%, TCI America™

Catalog No. p-7130113
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00421G 1 g
G00425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G00421G Supplier TCI America Supplier No. G00421G
Only null left

Chemical Identifiers

CAS 3646-68-2
Molecular Formula C6H13NO6
Molecular Weight (g/mol) 195.17
MDL Number MFCD00037764
InChI Key UFYKDFXCZBTLOO-UHFFFAOYNA-N
Synonym D-Glucosaminic Acid
PubChem CID 73563
ChEBI CHEBI:17784
IUPAC Name 2-amino-3,4,5,6-tetrahydroxyhexanoic acid
SMILES NC(C(O)C(O)C(O)CO)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 195.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucosamic Acid