missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-D-Glucan from barley for use with Total Dietary Fiber Control Kit, TDF-C10, MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages and General Analytical
Specifications
Specifications
| CAS | 9041-22-9 |
| Packaging | Poly Bottle |
| Linear Formula | (C6H10O5)n |
| Quantity | 1 g |
| MDL Number | MFCD00466918 |
| Physical Form | Lyophilized Powder |