missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%

CAS: 19046-60-7 | C6H11K2O9P | 336.32 g/mol

Supplier:  Thermo Scientific Chemicals 337035000

 View more versions of this product

Catalog No. AC337035000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

α-D-Galactose 1-phosphate dipotassium salt pentahydrate
19046-60-7
97.5
Crystalline Powder and/or Crystals
98%
MFCD00065475
(5% in water) Clear colorless
[K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
336.32
426.39
Authentic
(5% aq. soln.) clear, colorless
98%
Colorless to White
100.0
500 mg
C6H11K2O9P
alpha-d-galactose-1-phosphate dipotassium salt dihydrate
KCIDZIIHRGYJAE-YGFYJFDDSA-L
potassium;[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
131875454
98%
Glass bottle

Chemical Identifiers

19046-60-7
336.32
KCIDZIIHRGYJAE-YGFYJFDDSA-L
131875454
[K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
C6H11K2O9P
MFCD00065475
alpha-d-galactose-1-phosphate dipotassium salt dihydrate
potassium;[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

EINECSNumber : 242-783-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only