missing translation for 'onlineSavingsMsg'
Learn More

D-Galacto-D-mannan

Catalog No. AAJ66610MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66610MC 100 mg
AAJ66610ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66610MC Supplier Thermo Scientific Chemicals Supplier No. J66610MC

CAS: 11078-30-1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11078-30-1
MDL Number MFCD00146683
Synonym galactomannan, galactomannoglycan, d-galacto-d-mannane, 6-o-alpha-d-galactopyranosyl-4-o-beta-d-mannopyranosyl-beta-d-mannopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyoxan-2-yl methoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-2-yl methoxy tetrahydropyran-3,4,5-triol, 2r,3s,4s,5s,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-2-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy oxane-3,4,5-triol

Specifications

Color White
Quantity 100 mg
Formula Weight ∽200000
Physical Form Powder
Chemical Name or Material D-Galacto-D-mannan
EINECSNumber 234-299-6
RTECSNumber OP3805000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only