missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Fructose, 99%

Catalog No. p-4510118
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161355000 500 g Plastic bottle
AC161350010 1 kg Plastic bottle
AC161350025 2.5 kg Plastic bottle
AC161350100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC161355000 Supplier Thermo Scientific Chemicals Supplier No. 161355000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D(-)-Fructose, 99%, C6H12O6, CAS Number-57-48-7, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, arabino-hexulose, d--fructose, d---fructose, d-levulose, fructose, d, furucton, keto-d-fructose, krystar 300, sugar, fruit, 500g, Passes test, 98.5% min, 31,321, CHEBI:48095, 50ppm max, White, 200-333-3 | CAS: 57-48-7 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 103.0°C to 105.0°C
Color White
Loss on Drying 0.5% (70°C, 4 hrs) (vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Plastic bottle
Quantity 500 g
Beilstein 31,321
Merck Index 15,4302
Specific Rotation Condition − 92.25 (20.00°C c=10,H2O,on dry sub.)
Specific Rotation − 92.25
Solubility Information Solubility in water: 3750g/L (20°C). Other solubilities: soluble in alcohol,pyridine,ether,ethylamine,and methylamine
Formula Weight 180.16
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(-)-Fructose
Show More Show Less
EINECSNumber 200-333-3

RUO – Research Use Only