missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-6-phosphate disodium salt, 95%

Catalog No. AAJ66311ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66311ME 500 mg
AAJ66311MC 100 mg
AAJ6631103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66311ME Supplier Thermo Scientific Chemicals Supplier No. J66311ME

CAS: 26177-86-6 | C6H13Na2O9P | 306.11 g/mol

D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-6-phosphate disodium salt is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

Solubility
Soluble in water (100 mg/ml).

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 26177-86-6
Molecular Formula C6H13Na2O9P
Molecular Weight (g/mol) 306.11
MDL Number MFCD16661244
InChI Key VAPLWAFNCWDFQF-UHFFFAOYNA-N
PubChem CID 129893509
IUPAC Name sodium;[(2R,3R,4S)-2,3,4,6-tetrahydroxy-5-oxohexyl] dihydrogen phosphate
SMILES [Na].[Na].OCC(=O)C(O)C(O)C(O)COP(O)(O)=O

Specifications

Color White
Quantity 500 mg
Solubility Information Soluble in water (100mg/ml).
Formula Weight 304.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-Fructose-6-phosphate disodium salt
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 247-505-4
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only