missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose 1,6-bisphosphate trisodium salt octahydrate, 98%

CAS: 81028-91-3 | C6H26Na3O20P2 | 549.17 g/mol

Supplier:  Thermo Scientific Chemicals J6625622

 View more versions of this product

Catalog No. AAJ6625622

Only null left
Explore available promotions

Description

Description

D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

Solubility
Soluble in water

Notes
Moisture Sensitive, store away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

D-Fructose 1,6-bisphosphate trisodium salt octahydrate
White
MFCD00149340,MFCD00135870,MFCD00068321
3837985
FDP-Na3H
DBTJWGHAMYSWQR-OLMHPWOBSA-J
trisodium octahydrate [(3S,4S,5R)-2,3,4-trihydroxy-5-[(phosphonatooxy)methyl]oxolan-2-yl]methyl phosphate
550.18
Powder
81028-91-3
C6H26Na3O20P2
100 g
Moisture sensitive
Soluble in water
O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].O[C@H]1[C@H](O)C(O)(COP([O-])([O-])=O)O[C@@H]1COP([O-])([O-])=O
549.17
98%

Chemical Identifiers

81028-91-3
549.17
DBTJWGHAMYSWQR-OLMHPWOBSA-J
trisodium octahydrate [(3S,4S,5R)-2,3,4-trihydroxy-5-[(phosphonatooxy)methyl]oxolan-2-yl]methyl phosphate
C6H26Na3O20P2
MFCD00149340,MFCD00135870,MFCD00068321
FDP-Na3H
O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].O[C@H]1[C@H](O)C(O)(COP([O-])([O-])=O)O[C@@H]1COP([O-])([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 253-778-0

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only