Learn More
Thermo Scientific Chemicals D-Fructose 1,6-bisphosphate trisodium salt octahydrate, 98%
Description
D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.
Solubility
Soluble in water
Notes
Moisture Sensitive, store away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Color | White |
| Quantity | 10 g |
| Beilstein | 3837985 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water |
| Formula Weight | 550.18 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | D-Fructose 1,6-bisphosphate trisodium salt octahydrate |
Safety and Handling
| EINECSNumber | 253-778-0 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only