missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-1,6-bisphosphate trisodium salt

Catalog No. aaj6634903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6634903 1 g
AAJ6634906 5 g
AAJ6634914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6634903 Supplier Thermo Scientific Chemicals Supplier No. J6634903

CAS: 38099-82-0 | C6H11Na3O12P2 | 406.059 g/mol

D-Fructose-1,6-bisphosphate trisodium salt is used to reduce ischemia-reperfusion injury in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-bisphosphate trisodium salt is used to reduce ischemia-reperfusion injury in rats.

Solubility
Soluble in water at 50mg/ml

Notes
Protect from heat. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 38099-82-0
Molecular Formula C6H11Na3O12P2
Molecular Weight (g/mol) 406.059
MDL Number MFCD00135872
InChI Key CEHGLSKJDFICTB-PWVOXRODSA-K
Synonym unii-t1e52g2c9i, d-fructose-1,6-diphosphate trisodium salt, fructose-1,6-bis dihydrophosphate , trisodium salt, fosfructose trisodium anhydrous, trisodium 1-o-hydroxyphosphinato-6-o-phosphonato-d-fructose, trisodium 2r,3r,4s-2,3,4-trihydroxy-6-hydroxy oxido phosphoryl oxy-5-oxohexyl phosphate
PubChem CID 94482
IUPAC Name trisodium;[(2R,3R,4S)-2,3,4-trihydroxy-6-[hydroxy(oxido)phosphoryl]oxy-5-oxohexyl] phosphate
SMILES C(C(C(C(C(=O)COP(=O)(O)[O-])O)O)O)OP(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water at 50mg/ml
Formula Weight 406.06
Physical Form Powder
Chemical Name or Material D-Fructose-1,6-bisphosphate trisodium salt
EINECSNumber 253-778-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only