missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Erythronolactone 98.0+%, TCI America™
Supplier: TCI America E045525G
Specifications
| D-Erythronolactone | |
| 103°C | |
| C4H6O4 | |
| 25 g | |
| SGMJBNSHAZVGMC-PWNYCUMCSA-N | |
| (3R,4R)-3,4-dihydroxyoxolan-2-one | |
| 5325915 | |
| ≥98.0% (GC) |
| 15667-21-7 | |
| White | |
| MFCD00077763 | |
| d-erythronolactone, 3r,4r-3,4-dihydroxydihydrofuran-2 3h-one, erythrono-1,4-lactone, 3r,4r-3,4-dihydroxyoxolan-2-one, 3r,4r-3,4-dihydroxyoxolan-2-one, d-erythronic acid gamma-lactone, d-erythronic acid gamma-lactone, pubchem5782, 2r,3r-butane-2,3,4-triol-1,4 lactone, a-lactone, a-lactone | |
| C1C(C(C(=O)O1)O)O | |
| 118.088 | |
| 118.09 | |
| Crystalline Powder |
Chemical Identifiers
| 15667-21-7 | |
| 118.088 | |
| SGMJBNSHAZVGMC-PWNYCUMCSA-N | |
| 5325915 | |
| C1C(C(C(=O)O1)O)O |
| C4H6O4 | |
| MFCD00077763 | |
| d-erythronolactone, 3r,4r-3,4-dihydroxydihydrofuran-2 3h-one, erythrono-1,4-lactone, 3r,4r-3,4-dihydroxyoxolan-2-one, 3r,4r-3,4-dihydroxyoxolan-2-one, d-erythronic acid gamma-lactone, d-erythronic acid gamma-lactone, pubchem5782, 2r,3r-butane-2,3,4-triol-1,4 lactone, a-lactone, a-lactone | |
| (3R,4R)-3,4-dihydroxyoxolan-2-one |
Safety and Handling
TSCA : No