missing translation for 'onlineSavingsMsg'
Learn More

D-Erythronolactone 98.0+%, TCI America™

Catalog No. E04555G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04555G 5 g
E04551G 1 g
E045525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E04555G Supplier TCI America Supplier No. E04555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15667-21-7
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.088
MDL Number MFCD00077763
InChI Key SGMJBNSHAZVGMC-PWNYCUMCSA-N
Synonym d-erythronolactone, 3r,4r-3,4-dihydroxydihydrofuran-2 3h-one, erythrono-1,4-lactone, 3r,4r-3,4-dihydroxyoxolan-2-one, d-erythrono-1,4-lactone, d-erythronic acid gamma-lactone, pubchem5782, 2r,3r-butane-2,3,4-triol-1,4 lactone, d-erythronic, a-lactone, erythronic acid g-lactone
PubChem CID 5325915
IUPAC Name (3R,4R)-3,4-dihydroxyoxolan-2-one
SMILES C1C(C(C(=O)O1)O)O

Specifications

Melting Point 103°C
Color White
Quantity 5 g
Formula Weight 118.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-Erythronolactone